Learn More
Cole-Parmer Jenway 632 521 Visible Spectrophotometer with Domed Lid, 110 VAC
Supplier:  Cole-Parmer 8305400
Encompass
This easy-to-use spectrophotometer is ideal for the busy technician or student. Wavelength calibration is automatic at start-up and includes an auto-zero (% transmittance) functiona significant time saver. This spectrophotometer is fully GLP (Good Laboratory Practice) compliant. Each sample reading is given a time and date stamp and easily connects to a printerthen simply push the print key to send data. The included 63-Zero PC software allows you to take and control measurements remotely, and export data and results to Excel, Word, Power Point, and other software applications for storage, reporting, or presentation. Transmittance, absorbance, or concentration with factor value, as well as wavelength are simultaneously. The digital, dual-line LCD shows error messages, prompts, mode indication, and concentration units (ppm, mg/L, g/L, M, %, or blank).
Catalog No. 50-196-1209