Learn More
PCE AMERICAS INC Colorimeter measuring geometry 10° or 2°/d
SDP
Catalog No. NC3654621
Encompass_Preferred
"The color meter PCE-CSM 10 was developed for quality control, offers a high accuracy and comes with a 3.5"" touch screen display. When using the color meter, you can choose between different color spaces (CIE L*A*B, XYZ, Yxy, LCh, CIE LUV, HunterLAB). After a measurement with the color meter, the reading can be used as a reference value while other readings can be recorded as a sample and later be compared to the reference value easily. In addition to the individual color space coordinates, the deviation between the reading is also displayed. When using the color meter, it is possible to store up to 1000 reference values and up to 15,000 samples on the device. The measured data can also be transferred to a PC via the USB interface. A special PC software is included in the package."
Catalog No. NC3654621 Supplier PCE AMERICAS INC Supplier No. PCECSM 10
Add to Cart
Add to Cart
Request a Quote
"The color meter PCE-CSM 10 was developed for quality control, offers a high accuracy and comes with a 3.5"" touch screen display. When using the color meter, you can choose between different color spaces (CIE L*A*B, XYZ, Yxy, LCh, CIE LUV, HunterLAB). After a measurement with the color meter, the reading can be used as a reference value while other readings can be recorded as a sample and later be compared to the reference value easily. In addition to the individual color space coordinates, the deviation between the reading is also displayed. When using the color meter, it is possible to store up to 1000 reference values and up to 15,000 samples on the device. The measured data can also be transferred to a PC via the USB interface. A special PC software is included in the package."