Learn More
PCE AMERICAS INC Colorimeter featuring a measuring 45/0 according to CIE No. 15, GB/T3978
SDP
Catalog No. NC3654881
Encompass_Preferred
"The color meter and spectrophotometer PCE-CSM 8 was developed for quality control, offers a high accuracy and comes with a 3.5"" touch screen display. When using the color meter, you can choose between different color spaces (CIE LAB, XYZ, Yxy, LCh, and LUV) and 45/0 optics angle. After a measurement with the color meter, the reading can be used as a named reference value while other readings can be recorded as a sample and later be compared to the reference value easily for Pass/Fail determination. In addition to the individual color space coordinates, the deviation between the reading (delta E) is also displayed. When using the color meter, it is possible to store up to 1000 reference values and up to 10,000 samples on the device. The measured data can also be transferred to a PC via the USB interface. A special PC software is included in the package."
Catalog No. NC3654881 Supplier PCE AMERICAS INC Supplier No. PCECSM 8
Add to Cart
Add to Cart
Request a Quote
"The color meter and spectrophotometer PCE-CSM 8 was developed for quality control, offers a high accuracy and comes with a 3.5"" touch screen display. When using the color meter, you can choose between different color spaces (CIE LAB, XYZ, Yxy, LCh, and LUV) and 45/0 optics angle. After a measurement with the color meter, the reading can be used as a named reference value while other readings can be recorded as a sample and later be compared to the reference value easily for Pass/Fail determination. In addition to the individual color space coordinates, the deviation between the reading (delta E) is also displayed. When using the color meter, it is possible to store up to 1000 reference values and up to 10,000 samples on the device. The measured data can also be transferred to a PC via the USB interface. A special PC software is included in the package."