Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO Spectrophotometer ScanStart™ Easy Vis Water Testing Analyzer

Catalog No. 13687275 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Voltage:
100/240 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
13-687-275 100/240 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-687-275 Supplier METTLER TOLEDO Supplier No. 30997810
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easy VIS are highly reliable, rapid, and space-efficient spectrophotometers extensively used for precise water quality analysis. Together, these features make the system ideal for fast, flexible, and reliable testing of waste, drinking, and process water across various industrial sectors.

  • ScanStart™ bundles equipped with barcode scanners enable seamless access to over 80 common wastewater, drinking water, and process water analytes and support flexible operation with NANOCOLOR™ test kits from Macherey-Nagel
  • NANOCOLOR™ test kits greatly simplify sample preparation by including all necessary reagents, effectively eliminating the need for time-consuming and costly chemical preparation steps.
  • Designed for simplicity and ease of use, delivering highly accurate and consistent measurement results, as well as built-in validated methods for up to 30 color scales that automatically eliminate calculation and transcription errors.
  • Optical design is optimized to reduce effects of turbidity commonly found in wastewater samples
  • Avoid errors. PathDetect™ securely checks the path length to ensure accurate results
  • Enhanced usability features like a simplified display of concentration results and embedded performance tests significantly accelerate routine analyses.
  • EasyDirect Software enables data acquisition and management as well as export capability for ease of result access and review. An intuitive interface displays concentration results of pertinent analytes for EPA methodologies in water analysis
TRUSTED_SUSTAINABILITY

Specifications

Detector Type Dual Silicon Photodiodes
Display Touchscreen
Languages English, French, German, Chinese, Italian
Light Source Tungsten
Spectral Bandwidth 4 nm
Type VIS Spectrophotometer
Voltage 100/240 V
Wavelength Range 330 to 1000 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.