Learn More
IMPLEN INC Implen OD600 Spectrophotometer, Microbial Growth Analysis
Catalog No. NC3404474
Encompass_Preferred
Microbial Growth Analysis, The Implen OD600, is built for streamlined determination of cell density. Featuring two built-in programs, OD600 and McFarland, along with a sample-list-function for easy data management and export, this device offers unparalleled accuracy and convenience. The OD600 offers utmost compatibility with both cuvettes and tubes, without the need adapters. In addition to the user friendly design, the OD600 is linear up to 4A, providing high precision and versatile measurements. Onboard storage for over 500,000 measurements and export in xls or csv file types. The Implen OD600 is ideal for diverse laboratory environments, thanks to its user-friendly design, robust data management capabilities, and seamless LIMS integration. Track bacterial growth phases from lag to log and beyond with the Implen OD600!*Charger included; includes one pack of DiluCell™ 10 cuvettes (pack of 96).
Catalog No. NC3404474 Supplier IMPLEN INC Supplier No. OD600
Microbial Growth Analysis, The Implen OD600, is built for streamlined determination of cell density. Featuring two built-in programs, OD600 and McFarland, along with a sample-list-function for easy data management and export, this device offers unparalleled accuracy and convenience. The OD600 offers utmost compatibility with both cuvettes and tubes, without the need adapters. In addition to the user friendly design, the OD600 is linear up to 4A, providing high precision and versatile measurements. Onboard storage for over 500,000 measurements and export in xls or csv file types. The Implen OD600 is ideal for diverse laboratory environments, thanks to its user-friendly design, robust data management capabilities, and seamless LIMS integration. Track bacterial growth phases from lag to log and beyond with the Implen OD600!*Charger included; includes one pack of DiluCell™ 10 cuvettes (pack of 96).