Learn More
Thermo Scientific™ Spectrophotometer and UV/IR Cuvets
Description
Cuvets feature wide-range silica glass windows used in spectrophotometric measurements from 220 to 2500nm. Circular models measure 0.5 dia. x 4 in.L (13 x 101mm).
Specifications
Specifications
| Type | Test Tube Cuvette |
| Path Length | 12.7 mm |
| For Use With (Equipment) | Spectrophotometer |