Learn More
Starna Cells Inc SPECTROSIL FAC UV QUARTZ
SDP
Catalog No. NC9651589
Encompass_Preferred
Standard Rectangular Cells with Stopper Pathlength: 1mm Exterior Width: 12.5mm Exterior Length: 3.5mm Exterior Height: 55mm Interior Width: 10mm Interior Length: 1mm Nominal Volume: 0.400mL Number of Polished Windows: 2
Catalog No. NC9651589 Supplier Starna Cells Inc Supplier No. 21Q1
Standard Rectangular Cells with Stopper Pathlength: 1mm Exterior Width: 12.5mm Exterior Length: 3.5mm Exterior Height: 55mm Interior Width: 10mm Interior Length: 1mm Nominal Volume: 0.400mL Number of Polished Windows: 2