Learn More
Starna Cells Inc SPECTROSIL QUARTZ CUVETTE
SDP
Supplier:  Starna Cells Inc 28/9Q10
Encompass_Preferred
Spectrosil Quartz Cuvette, Path Length: 10mm, Exterior Size: 12.5L x 12.5W x 48mmH, Interior Size: 2W x 10mm long, Base Thickness: 9mm, Nominal Volume: 0.580 mL, Number Of polished Windows: 2
Catalog No. NC9030411