Learn More
Starna Cells Inc SPECTROSIL QUARTZ CUVETTE
SDP
Catalog No. NC9030412
Encompass_Preferred
Micro, w/stopper(s), Spectrosil quartz, 2 W Internal, 12.5L x 12.5W x 48H, 0.580mL, 9mm thick base, Self-masking, Black walls, Two polished windows, Walls polished internally, Fine ground externally
Catalog No. NC9030412 Supplier Starna Cells Inc Supplier No. 28B9Q10
Micro, w/stopper(s), Spectrosil quartz, 2 W Internal, 12.5L x 12.5W x 48H, 0.580mL, 9mm thick base, Self-masking, Black walls, Two polished windows, Walls polished internally, Fine ground externally