Learn More
Bolle Safety Spectrum Goggle
Catalog No. 17720233
Change view
Click to view available options
Frame Type:
Indirect Vented
Indirect Vented with Foam
Sealed
Sealed with Foam
Lens Tint:
Clear
Smoke
Model:
SPECTN10W
SPECTN11W
SPECTN12W
SPECTN13W
SPECTN14W
SPECTN15W
SPECTN20W
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Type Lens Tint Model
17-720-233 Indirect Vented Clear SPECTN10W
17-720-227 Indirect Vented Smoke SPECTN20W
17-720-228 Sealed Clear SPECTN15W
17-720-229 Indirect Vented Clear SPECTN14W
17-720-230 Sealed with Foam Clear SPECTN13W
17-720-231 Sealed Clear SPECTN12W
17-720-232 Indirect Vented with Foam Clear SPECTN11W
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-720-233 Supplier Bolle Safety Supplier No. SPECTN10W
Only null left
Add to cart
Add to cart
With its bi-material frame and adjustable strap, Spectrum offers superior protection and adaptability. Available in two sizes, Spectrum is a versatile product with different frame types and lens coatings to provide a solution for every company, regardless of their specific work environment. Its sporty design and optimized ergonomics also ensure greater acceptance for enhanced safety and productivity.
  • PC/TPR Co-injected frame for optimum comfort.
  • Multiple coatings and sizes available.
  • Soft gasket to fit most face structures.
  • Optimal panoramic vision.
Specifications
Lens Coating Platinum
Lens Tint Clear
Product Type Safety Goggles
Model SPECTN10W
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection None
Frame Type Indirect Vented
Autoclavable Not Autoclavable