Learn More
Spectrum™ Standard RC Ready-to-use Dialysis Sacks Spectra/Por™
SDP
Catalog No. 08671171 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
MWCO:
12 to 14 kDa
Catalog No. MWCO
08-671-171 12 to 14 kDa
1 options
Catalog No. 08-671-171 Supplier Spectrum™ Supplier No. 132666
Only null left
Add to Cart
Add to Cart
Specifications
Product Type Ready-to-Use Dialysis Sacks
MWCO 12 to 14 kDa
Membrane Spectra/Por 1
Width (Metric) 50 mm
Capacity (Metric) 50 to 400 mL