Learn More
Spectrum™ Trial Size Kits for Biotech-Grade CE Dialysis Membrane Tubing (1 meter), MWCO: 20 kd
Shop All Spectrum Medical Industries Products

Description
Includes
- 1m roll of Biotech CE Dialysis Tubing (16mm flatwidth)
- 2 Universal tubing closures
- 5 Tubing opening picks
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
Kits are great for comparing membrane MWCO performance to optimize separation or purification. Provides a 1m length of Biotech-Grade CE membrane tubing (16mm flat width), a pair of universal tubing closures and 5 picks for opening membrane tubing. Tubing is wet packaged in 0.05% sodium azide preservative that easily rinses out with water. Nine different MWCOs avilalable: 0.1-0.5 kD, 0.5-1.0 kD, 3.5-5 kD, 8-10 kD, 20 kD, 50 kD, 100 kD, 300 kD and 1000 kD.
- Rigidly controlled porosity for better selectivity and separations than Standard Grade RC
- 9 MWCOs available ranging from 100 to 1,000,000 Daltons
- Ultrapure, no heavy metal or sulfiide impurities
- Low protein binding
- No glycerin removal, cleaning or pre-treatment required
Specifications
Specifications
| Product Type | Dialysis Trial Kit |
| MWCO | 20 kDa |
| Format | Tubing |
| Diameter (Metric) | 10 mm |
| Diameter (English) | 0.39 in. |
| Kit Contents | 1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks |
| Quantity | Each |
| For Use With (Application) | Ideal for small, critical dialysis applications |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.