Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Microdialysis Hollow Fiber
SDP

Catalog No. 08670308 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
MWCO:
13 kDa
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
08-670-308 13 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-670-308 Supplier Spectrum™ Supplier No. 132294
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Product Type Microdialysis Hollow Fiber
MWCO 13 kDa
Membrane Regenerated Cellulose
Material Regenerated Cellulose
Length (English) 6 in.
Length (Metric) 152 mm
Capacity (Metric) 5 μL
Quantity Each
For Use With (Application) The implantation of individual micro-dialysis hollow fibers is an ideal step in the recovery or introduction of micro-quantity biologicals or pharmaceutical materials in a localized tissue environment
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.