Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Standard RC Ready-to-use Spectra/Por™ Dialysis Sacks
SDP

Catalog No. 08671172 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
MWCO:
12 to 14 kDa
3.5 kDa
Diameter (English):
1.14 in.
1.33 in.
Membrane:
Spectra/Por 2
Spectra/Por 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Diameter (English) Membrane
08-671-172 12 to 14 kDa 1.14 in. Spectra/Por 2
08-801-06 3.5 kDa 1.33 in. Spectra/Por 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-671-172 Supplier Spectrum™ Supplier No. 132681
Add to Cart
Add to Cart

For large sample volumes

  • Material: Regenerated Cellulose
  • Length: 60cm (23.62 in.)
  • Sealed at one end, open at the other
  • Funnel attached to top for easy sample filling
  • Supplied in sodium azide 0.05% (must be rinsed prior to use)

Specifications

Product Type Dialysis Sacks
MWCO 12 to 14 kDa
Membrane Spectra/Por 2
Material Regenerated Cellulose
Diameter (Metric) 29 mm
Diameter (English) 1.14 in.
Length (English) 23.6 in.
Length (Metric) 600 mm
Width (Metric) 45 mm
Capacity (Metric) 50 to 400 mL
Kit Contents 0.05% sodium azide preservant
Quantity 10 Pack
For Use With (Application) For large sample volumes, funnel attached to top for easy sample filling
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.