Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Spectrum Tube
SCI_ED

Catalog No. p-7229339 Shop All Supertek Scientific Products
Change view
Click to view available options
Product Type:
Spectrum Tube (Argon Gas)
Spectrum Tube (Hydrogen Gas)
Spectrum Tube (Krypton Gas)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S139447 Spectrum Tube (Krypton Gas)
S139448 Spectrum Tube (Argon Gas)
S139449 Spectrum Tube (Hydrogen Gas)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S139447 Supplier Supertek™ Supplier No. PH95014/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

The spectrum tube demonstrates electrons in the gas to be excited as an electrical discharge passes through the tube. As the electrons relax, they emit light-a characteristic color for each gas.

  • Tubes are 26 cm long overall narrowed to capillary width for 8.5 to 10 cm of the length.
  • Metal wires holding the electrodes are sealed through the ends and welded to metal caps which have loops for connecting wires.

Specifications

Grade 9+
Format Kit
Product Type Spectrum Tube (Krypton Gas)
Material Glass
Languages English
Subject Physical Science - Gases
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.