Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Spectrum Tubes
SDPSCI_ED

Catalog No. S44002F Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Argon Spectrum Tube
Krypton Spectrum Tube
Neon Spectrum Tube
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S44002F Krypton Spectrum Tube
S43993F Neon Spectrum Tube
S43991F Argon Spectrum Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S44002F

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Discharge apparatus such as spectrum tube power supply, static machine or induction coil

Reveal the spectra of various gases when discharges pass through them.

Provide a visual representation of the energy associated with electron transitions within excited atoms and molecules.

  • Made from two glass bulbs connected by a capillary tube
  • Rod electrodes at each end have cap and ring terminals
  • Small cross-sectional area causes the discharge to produce an especially radiant glow in the capillary
  • Wavelength may be determined with a diffraction grating spectrometer (S63131 or S42592)
  • Length: 10.5 in. (266mm)

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
For Use With (Application) Provides a visual representation of the energy associated with electron transitions within excited atoms and molecules
For Use With (Equipment) Spectrometer
Material Glass
Length (English) 10.5 in.
Length (Metric) 26.7 cm
Product Type Krypton Spectrum Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.