Learn More
Perkin Elmer US LLC Spectrum Two Battery Pack
Catalog No. 502117232
Encompass_Preferred
The Spectrum Two Battery Pack Battery Pack is an optional power supply for the Spectrum Two. This uninterruptable power source is an 18 V battery with a maximum current of 3 amps. The batter pack includes a 12 V USB car auxiliary port adapter. Required, but not included, is a voltage-specific, country power cord.
Catalog No. 50-211-7232 Supplier Perkin Elmer US LLC Supplier No. L1600233
The Spectrum Two Battery Pack Battery Pack is an optional power supply for the Spectrum Two. This uninterruptable power source is an 18 V battery with a maximum current of 3 amps. The batter pack includes a 12 V USB car auxiliary port adapter. Required, but not included, is a voltage-specific, country power cord.