Learn More
Med Vet International Miltex Meisterhand Barraquer Eye Speculum, Small, 1.75in, German, Mh18-36
Catalog No. 502831500
Encompass_Preferred
Commonly used ophthalmologically when visualization of the globe is needed. Both end pieces of the blade can be fit into the upper and lower lid. Solid blades measuring out to 10mm. 1.75 in total. Small. MeisterHand build quality ensures superior craftsmanship. Made with high quality German steel.
Catalog No. 50-283-1500 Supplier Med Vet International Supplier No. MILMH1836
Commonly used ophthalmologically when visualization of the globe is needed. Both end pieces of the blade can be fit into the upper and lower lid. Solid blades measuring out to 10mm. 1.75 in total. Small. MeisterHand build quality ensures superior craftsmanship. Made with high quality German steel.