Learn More
Med Vet International Miltex Graves Vaginal Speculum, Original, Small, 0.75in X 3in, German, 30-10
Catalog No. 502832442
Encompass_Preferred
Open sided single-end vaginal speculum commonly used in OB/GYN procedures such as vaginal hysterectomies or dilation. Small. Wide angle blade. 6.75in total length. 0.75in X 3in. Made with high quality German steel.
Catalog No. 50-283-2442 Supplier Med Vet International Supplier No. MIL3010
Open sided single-end vaginal speculum commonly used in OB/GYN procedures such as vaginal hysterectomies or dilation. Small. Wide angle blade. 6.75in total length. 0.75in X 3in. Made with high quality German steel.