Learn More
Med Vet International Miltex Graves Vaginal Speculum, Side Screw w/Nut, German, 30-15A
Catalog No. 502832097
Encompass_Preferred
Open sided single-end vaginal speculum commonly used in OB/GYN procedures such as vaginal hysterectomies or dilation. Side screw allow the practitioner to lock the jaws into place letting them free their hand up for more extensive work. Made with high quality German steel.
Catalog No. 50-283-2097 Supplier Med Vet International Supplier No. MIL3015A
Open sided single-end vaginal speculum commonly used in OB/GYN procedures such as vaginal hysterectomies or dilation. Side screw allow the practitioner to lock the jaws into place letting them free their hand up for more extensive work. Made with high quality German steel.