Learn More
Med Vet International Miltex Knapp Eye Speculum, 3.12in, 15mm Blades, German, 18-4
Catalog No. 502829091
Encompass_Preferred
Instrument frequently used in visualizing the curvature of the eye. The speculum is designed to accommodate up to a 60.0 mm spread with a locking screw to hold in place. 3.12in. 15mm blades. Made with high quality German steel.
Catalog No. 50-282-9091 Supplier Med Vet International Supplier No. MIL184
Instrument frequently used in visualizing the curvature of the eye. The speculum is designed to accommodate up to a 60.0 mm spread with a locking screw to hold in place. 3.12in. 15mm blades. Made with high quality German steel.