Learn More
Med Vet International Miltex Lancaster Eye Speculum, 15.5mm Blades, 3.12in, German, 18-2
Catalog No. 502829095
Encompass_Preferred
Used to visualize the curvature of the eye. Adult size, 15.5mm round solid blades. 3.12in. Made with high quality German steel.
Catalog No. 50-282-9095 Supplier Med Vet International Supplier No. MIL182
Used to visualize the curvature of the eye. Adult size, 15.5mm round solid blades. 3.12in. Made with high quality German steel.