Learn More
Med Vet International Miltex Mellinger Eye Speculum, Spread, 1.12in, German, 18-66
Catalog No. 502830498
Encompass_Preferred
Commonly used ophthalmologically when visualization of the globe is needed. Both end pieces of the blade can be fit into the upper and lower lid. Spread solid blades measuring out to 15mm. 1.12 in total. Small. MeisterHand build quality ensures superior craftsmanship. Made with high quality German steel.
Catalog No. 50-283-0498 Supplier Med Vet International Supplier No. MIL1866
Commonly used ophthalmologically when visualization of the globe is needed. Both end pieces of the blade can be fit into the upper and lower lid. Spread solid blades measuring out to 15mm. 1.12 in total. Small. MeisterHand build quality ensures superior craftsmanship. Made with high quality German steel.