Learn More
Med Vet International Miltex Plain Wire Eye Speculum, 2.5in, 15mm Blades, German, 18-34
Catalog No. 502829088
Encompass_Preferred
Commonly used ophthalmologically when visualization of the globe is needed. Both end pieces of the blade can be fit into the upper and lower lid. Solid blades measuring out to 15mm. 2.5 in total. MeisterHand build quality ensures superior craftsmanship. Made with high quality German steel.
Catalog No. 50-282-9088 Supplier Med Vet International Supplier No. MIL1834
Commonly used ophthalmologically when visualization of the globe is needed. Both end pieces of the blade can be fit into the upper and lower lid. Solid blades measuring out to 15mm. 2.5 in total. MeisterHand build quality ensures superior craftsmanship. Made with high quality German steel.