Learn More
Med Vet International Miltex MeisterHand Rectal Speculum, Fenestrated Blades, 6in, German, MH28-2
Catalog No. 503047369
Encompass_Preferred
Speculum Designed for the dilation of the rectal cavity. Useful for specific procedures. MeisterHand build quality ensures superior craftsmanship. 7.5in long. Blades are 3.25in X 0.62in. Fenestrated blades. Wide at distal end. Made with High Quality German steel.
Catalog No. 50-304-7369 Supplier Med Vet International Supplier No. MILMH282
Speculum Designed for the dilation of the rectal cavity. Useful for specific procedures. MeisterHand build quality ensures superior craftsmanship. 7.5in long. Blades are 3.25in X 0.62in. Fenestrated blades. Wide at distal end. Made with High Quality German steel.