Learn More
Med Vet International Miltex Vienna Nasal Speculum, Medium, 5.25in, German, 20-3
Catalog No. 502829347
Encompass_Preferred
Tool that is useful for retracting nasal walls and inspecting nasal passageways. The double-spring design provides the perfect amount of tension when dilating. Medium build. Child. 5.25in. 29.5mm deep. Made with high quality German steel.
Catalog No. 50-282-9347 Supplier Med Vet International Supplier No. MIL203
Tool that is useful for retracting nasal walls and inspecting nasal passageways. The double-spring design provides the perfect amount of tension when dilating. Medium build. Child. 5.25in. 29.5mm deep. Made with high quality German steel.