Learn More
3B Scientific™ Speed of Light Meter
SDPSCI_ED
Catalog No. S97703 Shop All 3B Scientific Products
Change view
Click to view available options
For Use With (Application):
For determining the speed of light
Catalog No. For Use With (Application)
S97703 For determining the speed of light
1 options
Catalog No. S97703 Supplier 3B Scientific™ Supplier No. U8476460115
Add to Cart
Add to Cart
Includes
Basic unit including the oscillator, receiver, and built-in mains supply; shaft-mounted fresnel lens; and shaft-mounted triple-prism reflector
Determine the speed of light using electronic time-delay measurement
  • Compact housing holds the oscillator for the LED light pulse, the photoreceptor, and the calibrated quartz-crystal oscillator, which generates accurately timed square-wave pulses
  • Internal and external reflectors transmit a signal superimposed onto the original signal
  • The external main reflector features numerous, small triple-prism reflectors and can be set up by the naked eye
  • The evaluation is carried out with a dual-channel oscilloscope capable of recording at least 70 MHz (sold separately)
  • Speed of light can be calculated based on the determined differential delay
Order Info
Electrical Requirements: 115V, 50/60 Hz
Specifications
Product Type Speed of Light Meter
For Use With (Application) For determining the speed of light
Includes Basic unit including the oscillator, receiver and built-in mains supply, shaft-mounted fresnel lens and shaft-mounted triple-prism reflector