Learn More
Grainger 1/4 in General Purpose Air Regulator , 15 cfm Max. Flow - Regulators
Catalog No. NC1779081
Encompass_Preferred
Features a modular connector that allows easy removal and swap out, plus 2 full-flow gauge ports. Manifold regulator provides multiple pressures from a common inlet. For Max. Flow, the CFM flow is measured with 150 psig inlet adjusted to set pressure of 90 psig (while increasing flow) and a 15 psig drop.
Catalog No. NC1779081 Supplier Grainger Supplier No. 4ZM08
Features a modular connector that allows easy removal and swap out, plus 2 full-flow gauge ports. Manifold regulator provides multiple pressures from a common inlet. For Max. Flow, the CFM flow is measured with 150 psig inlet adjusted to set pressure of 90 psig (while increasing flow) and a 15 psig drop.