Learn More
Grainger SPEEDAIRE Filter Regulator, 1/4 in NPT
Supplier:  Grainger 4ZK76
Encompass_Preferred
The Speedaire 1/4 in. NPT Filter Regulator is a combination filter and pressure regulator designed to remove dirt, pipe scale, and water from compressed air in industrial compressed air systems. It features a die-cast aluminum body construction with 1/4 in. NPT inlet and outlet ports. The filter element is rated for 40 micron filtration. The pressure regulator is adjustable from 0-125 psi. This filter regulator unit provides both particle removal and pressure regulation in one compact unit. It is suitable for point-of-use installation in industrial compressed air systems.
Catalog No. NC2390264