Learn More
MSC Betco All-Purpose Cleaner, Concentrate, Melon & Mango Fragrance
Catalog No. 502837491
Encompass
Top Flite All-Purpose Cleaner is a concentrate designed to clean and deodorize most water-washable surfaces. Its advanced formula is made without butyl or inorganic phosphates, ensuring it will not harm most surfaces. It is suitable for mop, bucket, spray, or sponge applications and features a fresh Melon & Mango fragrance.
  • Suspends the toughest soils and prevents redepositing.
  • Cleans and deodorizes practically any water washable surface.
  • Extremely effective in hard water.
Catalog No. 50-283-7491 Supplier MSC Supplier No. 72969751
Top Flite All-Purpose Cleaner is a concentrate designed to clean and deodorize most water-washable surfaces. Its advanced formula is made without butyl or inorganic phosphates, ensuring it will not harm most surfaces. It is suitable for mop, bucket, spray, or sponge applications and features a fresh Melon & Mango fragrance.
  • Suspends the toughest soils and prevents redepositing.
  • Cleans and deodorizes practically any water washable surface.
  • Extremely effective in hard water.