Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ 9100 FX-Air Replacement Spring

Catalog No. 19071119 Shop All 3M Products
Change view
Click to view available options
Quantity:
2/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-071-119 2/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-071-119 Supplier 3M™ Supplier No. 7100215111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spring mechanism is a replaceable spare part for the 3M™ Speedglas™ Welding Helmet

  • For use with 9100 FX and 9100 FX-Air Welding Helmets
  • User can easily install
  • 3M Catalog# 06-0400-56

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) 9100 FX Welding Helmets and 9100 FX-Air Welding Helmets
For Use With (Application) Assembly, Grinding, Machine operations, sanding, welding
Model 06-0400-56
Product Type Spring
Quantity 2/Bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.