Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Airduct Breathing Tube with Holder

Catalog No. 19072128 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Speedglas™ Heavy-Duty Welding Helmet G5-01
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-072-128 Speedglas™ Heavy-Duty Welding Helmet G5-01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-128 Supplier 3M™ Supplier No. 7100215556
Add to Cart
Add to Cart

Spare part for 3M™ Speedglas™ Heavy-Duty Welding Helmet G5-01

  • To be ordered if the air duct breathing tube and/or holder has become damaged and needs replacement
  • For maximum safety and comfort, make it a habit to check equipment regularly
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Breathing Tube
For Use With (Application) Metal Repair, metal processing, welding, grinding
For Use With (Equipment) Speedglas™ Heavy-Duty Welding Helmet G5-01
Includes Holder
Model G5-01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.