Learn More
3M™ Speedglas™ FA III SAR V-100 and Speedglas™ Welding Helmet
Catalog No. 19071109 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-071-109 1 Ea.
1 options
Catalog No. 19-071-109 Supplier 3M™ Supplier No. 7100194538
Add to Cart
Add to Cart
Front of face air delivery helps provide cooling for hot work environments, while keeping the grinding visor virtually fog free
  • Smooth flip-up pivot mechanism assures effortless transition from welding to grinding modes.
  • Offers Natural Color technology for better control of the weld puddle, greater precision, simplified workflow.
  • Switching gears is easy with a helmet that’s up for multiple tasks. With integrated protection offered by 3M PAPR systems, workers can go from welding to grinding without removing, locating or foregoing their PPE.
  • Various components of a 3M PAPR system are designed to work together as a single unit – combining respiratory, eye, face and in certain cases head protection that can be customized for almost any work environment.
  • Reduced risk of eye injury
  • Options for limited facial hair
  • Easier, more comfortable breathing
  • Includes V-100 Vortex air-cooling valve, belt, breathing tube, and complete 9100 FX-Air helmet assembly including 9100 XXi Auto Darkening Filter (ADF)
  • Flip front welding shield allows for a clear, wide-view protective visor for grinding and setup. Lightweight valve can lower temperature of air entering headpiece by up to 50°F (28°C).
  • 3M Catalog# 26-5702-30ISW
TRUSTED_SUSTAINABILITY
Specifications
Battery Life ADF: up to 1800 hours
Battery Type ADF: CR2032
Certifications/Compliance ANSI Z87.1-2010, NIOSH Approved
For Use With (Application) Assembly and Mechanical, Grinding, Metal repair, Welding
Includes Carry Bag, Supplied Air Valve, Breathing Tube, Welding Helmet, ADF
Model 26-5702-30ISW
Product Type Safety Helmet
Quantity 1 Ea.
Visor Type Clear Visor and Auto-Darkening Welding Lens