Learn More
3M™ Speedglas™ FA III SAR V-100 Valve and Speedglas™ Welding Helmet
Catalog No. 19071110 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-071-110 1 Ea.
1 options
Catalog No. 19-071-110 Supplier 3M™ Supplier No. 7100194537
9100-Air is designed for enhanced comfort and protection.
  • Helmet has a smooth up-and-down pivot action that gently locks in the ‘up’ position.
  • Comes with a highly adjustable head suspension system and a well-balanced design.
  • Switching gears is easy with a helmet that’s up for multiple tasks. With integrated protection offered by 3M PAPR systems, workers can go from welding to grinding without removing, locating or foregoing their PPE.
  • Various components of a 3M PAPR system are designed to work together as a single unit – combining respiratory, eye, face and in certain cases head protection that can be customized for almost any work environment.
  • Reduced risk of eye injury
  • Options for limited facial hair
  • Easier, more comfortable breathing
  • Includes V-100 Vortex air-cooling valve, belt, breathing tube, and complete 9100-Air helmet assembly
  • ADF sold separately
  • Lightweight belt-mounted valve permits welders to lower temperature of air entering headpiece by up to 50°F (28°C); designed for hot, strenuous jobs
  • Side windows - Shade 5 side filters that increase peripheral vision.
  • 3M Catalog# 25-5702-00SW
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010, NIOSH Approved
For Use With (Application) Assembly and Mechanical; Grinding; Metal Repair; Welding
Includes Carry Bag, Supplied Air Valve, Breathing Tube, Welding Helmet
Model 25-5702-00SW
Product Type Supplied Air (SA) Respiratory System
Quantity 1 Ea.
Visor Type Auto-Darkening Filter