Learn More
3M™ Speedglas™ FA III SAR V-100 Valve and Speedglas™ Welding Helmet with ADF
Catalog No. 19071111 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-071-111 1 Ea.
1 options
Catalog No. 19-071-111 Supplier 3M™ Supplier No. 7100194535
Add to Cart
Add to Cart
3M™ Speedglas™ Welding Helmet 9100 MP (Multi-Protection) seamlessly integrates 5 levels of welder protection: head, eye, face, respiratory and hearing into one extraordinarily comfortable and easy-to-use system.
  • The hard hat shell is made from heat resistant polycarbonate and meets the ANSI Z89.1-2009 requirements for Type 1 Class G hard hats.
  • The 9100 XXi ADF offers Natural Color technology for better control of the weld puddle, greater precision, simplified workflow.
  • Switching gears is easy with a helmet that’s up for multiple tasks. With integrated protection offered by 3M PAPR systems, workers can go from welding to grinding without removing, locating or foregoing their PPE.
  • The various components of a 3M PAPR system are designed to work together as a single unit – combining respiratory, eye, face and in certain cases head protection that can be customized for almost any work environment.
  • Reduced risk of eye injury
  • Options for limited facial hair
  • A wide (approx. 8in x 4.25in), curved, protective visor with excellent views up, down and peripherally. Meets the requirements of CSA Z94.3 and the high impact requirements of ANSI Z87.1-2010.
  • 3M Catalog# 27-5702-30ISW
TRUSTED_SUSTAINABILITY
Specifications
Battery Life ADF: up to 1800 hours
Battery Type ADF: CR2032
Certifications/Compliance ANSI Z89.1 Type I, Class G; ANSI Z87.1-2010; NIOSH Approved
For Use With (Application) Assembly and Mechanical, Welding, Metal Repair, Grinding
Includes Carry Bag, Supplied Air Valve, Breathing Tube, Welding Helmet, ADF
Model 27-5702-30ISW
Product Type Safety Helmet
Quantity 1 Ea.
Visor Type Clear Visor and Auto-Darkening Welding Lens