Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ G5 Series Carry and Storage Bag

Catalog No. 19072150 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Helmet and PAPR or SAR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-072-150 Helmet and PAPR or SAR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-150 Supplier 3M™ Supplier No. 7100215530
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

G5 Series carry and storage bag contains removable straps so the bag can be used as either a carry bag or backpack.

  • To protect welding respiratory system from scratches
  • Fits both helmet and powered respirator or supplied air regulator
TRUSTED_SUSTAINABILITY

Specifications

Product Type Storage Bag
Color Black
Model SG-95
Includes Removable Straps
For Use With (Application) Grinding, Welding, Metal Repair, Metal Processing
For Use With (Equipment) Helmet and PAPR or SAR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.