Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC SPEEDGLAS HEADBAND

Catalog No. 5000675065
Encompass

This product is a replacement headband designed for the 3M Speedglas 9100 series welding helmets.

  • Accessory type: headbands
  • For use with: 9100 series welding helmets
  • Color: black
  • Standards: ANSI Z87.1-2010
  • Series: 9100 FX-Air
  • Country of origin: Sweden
  • Headband replacement part

Catalog No. 50-006-75065 Supplier MSC Supplier No. 69953347
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a replacement headband designed for the 3M Speedglas 9100 series welding helmets.

  • Accessory type: headbands
  • For use with: 9100 series welding helmets
  • Color: black
  • Standards: ANSI Z87.1-2010
  • Series: 9100 FX-Air
  • Country of origin: Sweden
  • Headband replacement part

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.