Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Leather Head Cover

Catalog No. 19072164 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-164 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-164 Supplier 3M™ Supplier No. 7100215541
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Speedglas™ Leather Head Cover is made in rugged leather for more long term protection against sparks and spatter.

  • A variety of accessories available to help maximize protection and comfort.
  • For maximum safety and comfort, make it a habit to check equipment regularly.
  • Replace worn out or damaged parts.
TRUSTED_SUSTAINABILITY

Specifications

Description Flame retardant
Color Brown
Material Leather
Product Type Headcover
For Use With (Equipment) Speedglas™ Heavy-Duty Welding Helmet G5-01
Model 46-0700-84
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.