Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Task Light Pouch for Adflo™ Battery

Catalog No. 19072161 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M™ Speedglas™ Task Light
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-072-161 3M™ Speedglas™ Task Light
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-161 Supplier 3M™ Supplier No. 7100216922
Add to Cart
Add to Cart

Speedglas™ Task Light Pouch is made in flame-retardant fabric for protection against sparks and spatter.

  • Designed to be worn on belt.
  • For maximum safety and comfort, make it a habit to check equipment regularly
  • Replace worn out or damaged parts.
    TRUSTED_SUSTAINABILITY

    Specifications

    Color Black
    Product Type Battery Pouch
    For Use With (Application) Metal Repair, Welding, Grinding, Metal Processing
    For Use With (Equipment) 3M™ Speedglas™ Task Light
    Model 35-1099-00
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.