Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Welding Lens 9100 Replacement Lens

Catalog No. 19071118 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Passive shade 10
Passive shade 11
Passive shade 12
Passive shade 13
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-071-118 Passive shade 13
19-071-115 Passive shade 10
19-071-116 Passive shade 11
19-071-117 Passive shade 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-071-118 Supplier 3M™ Supplier No. 7010342412
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement lens only (Welding filter adaptor not included)

  • 3M Catalog# 06-0005-P10, 06-0005-P11, 06-0005-P12, 06-0005-P13

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) 9100 Series Welding Helmets
For Use With (Application) Assembly, Facility Maintenance, Grinding, Sanding, Welding
Lens Tint Passive shade 13
Model 06-0005-P13
Product Type Welding Lens
Quantity 2/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.