Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Welding Lens Assembly Passive Shade

Catalog No. 19071114 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-071-114 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-071-114 Supplier 3M™ Supplier No. 7010342411
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Passive welding filters can work well for welders who perform long continuous welds rather than tack welds or for use in wet environments.

  • Full Passive Lens Assembly
  • Excellent lower-cost lens option for large, 2.8 x 4.2 in. (approx.) viewing area.
  • 3M Catalog# 06-0000-P10

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) 9100 Series Welding Helmets
For Use With (Application) Assembly, Facility Maintenance, Grinding, Sanding, Welding
Model 06-0000-P10
Product Type Welding Lens Assembly
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.