Learn More
Thermo Scientific Savant™ SpeedVac™ Rotors for 15mL conical
Supplier:  Thermo Scientific RH5215
Encompass_Preferred
Centrifuge Tubes: 52 x 15mL conical, Large capacity models. For Use With SPD210, SPD2030, SPD300DDA (requires RAD200).
Catalog No. NC1741322