Learn More
Electron Microscopy Sciences Speedy Foil, 4 x 4"
SDP
Catalog No. 501926853
Encompass_Preferred
Pharmaceutical grade, pre-cut, interleaved aluminum foil squares. Time saving product that eliminates the wasteful practice of trimming pieces from over-sized rolls. Each size ships in its own ideally sized dispensing box that ensures squares remain crisp and flat. Squares are pre-cut to fit common laboratory container openings saving time. Ideal for beakers up to 500ml.
Size: 4 x 4"
Catalog No. 50-192-6853 Supplier Electron Microscopy Sciences Supplier No. 6040914
Pharmaceutical grade, pre-cut, interleaved aluminum foil squares. Time saving product that eliminates the wasteful practice of trimming pieces from over-sized rolls. Each size ships in its own ideally sized dispensing box that ensures squares remain crisp and flat. Squares are pre-cut to fit common laboratory container openings saving time. Ideal for beakers up to 500ml.
Size: 4 x 4"