Learn More
Comecerbrand Speedy Glove - Automatic Glove Tester for Integrity Testing - Stand Alone - US (912 MHz)
Catalog No. 17980815
Change view
Click to view available options
Product Type:
Glove Integrity Tester
Catalog No. Product Type
17-980-815 Glove Integrity Tester
1 options
Catalog No. 17-980-815 Supplier Comecerbrand Supplier No. NL0154000002
Add to Cart
Add to Cart
Request a Quote
Stand-alone glove integrity tester designed for automated checks and simple data transfer, supplied with docking station and RFID tags. BASE CONFIGURATION.
  • Battery-powered portable device for leak testing gloves installed on isolators or RABS where strict separation between the isolated area and external environment is required; performs pressure deterioration integrity tests compliant with ISO 14644-7 Annex E.5 and GMP Annex 1.
  • Suitable for isolators with glove flanges used in aseptic and containment processes, API and HAPI handling, and radiopharmaceutical applications.
  • Flexible adapter system allows testing on glove flanges currently on the market or any custom variants, with no restrictions on flange type.
  • Stand-alone US (912 MHz) system with USB-C data transfer, supplied with docking station and 20 programmed RFID glove port tags; glove port connection adapter not included and must be selected separately.
Specifications
Product Type Glove Integrity Tester