Learn More
Comecerbrand SpeedyGlove Calibration Flange
Catalog No. 17980820
Change view
Click to view available options
Product Type:
Flange
Catalog No. Product Type
17-980-820 Flange
1 options
Catalog No. 17-980-820 Supplier Comecerbrand Supplier No. NL0254020001
Add to Cart
Add to Cart
Request a Quote
Closed calibration flange used to mount the SpeedyGlove for leak-tightness checks during testing. OPTION.
  • Closed calibration flange used to mount the SpeedyGlove for leak-tightness checks.
  • Required to verify the leak tightness of the SpeedyGlove.
  • Allows pressure measurement when used with a calibrated pressure controller (not included).
  • Supports accurate and reliable calibration
Specifications
For Use With (Equipment) Glove Integrity Tester
Product Type Flange