Learn More
Comecerbrand SpeedyGlove Custom Connection
Catalog No. 17980814
Change view
Click to view available options
Product Type:
Flange
Catalog No. Product Type
17-980-814 Flange
1 options
Catalog No. 17-980-814 Supplier Comecerbrand Supplier No. CTB046
Add to Cart
Add to Cart
Request a Quote
Custom connection designed to adapt the SpeedyGlove unit to different gloveport configurations.
  • Custom connection adapters made to fit any gloveport.
  • Created based on a provided sample, drawing, 3-D model, or similar reference.
  • Ensures proper compatibility with the specific gloveport design.
  • Supports accurate and reliable connection for SpeedyGlove use.
Specifications
For Use With (Equipment) Glove Integrity Tester
Product Type Flange