Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Comecerbrand SpeedyGlove Suitcase, for Easy Transportation of SpeedyGlove

Catalog No. 17980822
Change view
Click to view available options
Product Type:
Storage Suitcase
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-980-822 Storage Suitcase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-980-822 Supplier Comecerbrand Supplier No. NL0254020003
Add to Cart
Add to Cart

Suitcase designed to provide easy and safe transportation of the SpeedyGlove unit. ACCESSORY.

  • SpeedyGlove Suitcase, for easy transportation of SpeedyGlove

Specifications

For Use With (Equipment) Glove Integrity Tester
Product Type Storage Suitcase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.