Learn More
Biotage SPEFRT PEFRITS 10U/25ML 100
SDP
Catalog No. 502311376
Encompass
Frit; Biotage; Isolute; For SPE; Polyethylene; 10um; 20mm (0.812 in.) dia.; Column type E, 25mL; 100/pack
Catalog No. 50-231-1376 Supplier Biotage Supplier No. 1201066E
Frit; Biotage; Isolute; For SPE; Polyethylene; 10um; 20mm (0.812 in.) dia.; Column type E, 25mL; 100/pack