Learn More
Biotage SPEFRT PEFRITS 10U/3ML 100
SDP
Catalog No. 502311372
Encompass
Frit; Biotage; Isolute; For SPE; Polyethylene; 10um; 9mm (0.375 in.) dia.; Column type B, 3mL; 100/pack
Catalog No. 50-231-1372 Supplier Biotage Supplier No. 1201062B
Frit; Biotage; Isolute; For SPE; Polyethylene; 10um; 9mm (0.375 in.) dia.; Column type B, 3mL; 100/pack