Learn More
Biotage SPEFRT PEFRITS 20U/150ML 50
SDP
Catalog No. 502311363
Encompass
Frit; Biotage; Isolute; For SPE; Polyethylene; 20um; 38mm (1.5 in.) dia.; Column type J, 150mL; 100/pack
Catalog No. 50-231-1363 Supplier Biotage Supplier No. 1201039J
Frit; Biotage; Isolute; For SPE; Polyethylene; 20um; 38mm (1.5 in.) dia.; Column type J, 150mL; 100/pack